Assay Details
| Gene Symbol: UBE2V2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034042.2 | NP_001029214.1 | 2 - 3 | 175 | 78 | |
| GenBank mRNA | AB098920.1 | - | 1 - 2 | 159 | 78 | |
| BC103159.1 | - | 2 - 3 | 176 | 78 | ||
More Information
Set Membership: |
|

UBE2V2
Genomic Map