Assay Details
| Gene Symbol: GULO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034043.2 | NP_001029215.1 | 4 - 5 | 439 | 65 | |
| GenBank mRNA | BC102936.1 | - | 4 - 5 | 440 | 65 | |
More Information
Set Membership: |
|

GULO
Genomic Map