Assay Details
| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034051.2 | NP_001029223.1 | 1 - 2 | 31 | 64 | |
| GenBank mRNA | BC102313.1 | - | 1 - 2 | 33 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102313.1, NM_001034051.2 transcripts.
Set Membership: |
|

RPL27
Genomic Map