Assay Details
| Gene Symbol: PGAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034054.1 | NP_001029226.1 | 2 - 3 | 420 | 73 | |
| GenBank mRNA | BC103115.1 | - | 2 - 3 | 440 | 73 | |
More Information
Set Membership: |
|

PGAM1
Genomic Map