Assay Details
| Gene Symbol: UBL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034214.2 | NP_001029386.1 | 1 | 525 | 61 | |
| GenBank mRNA | BC103189.1 | - | 1 | 526 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBL4A| Gene Symbol: UBL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034214.2 | NP_001029386.1 | 1 | 525 | 61 | |
| GenBank mRNA | BC103189.1 | - | 1 | 526 | 61 | |
Set Membership: |
|