Assay Details
| Gene Symbol: EIF4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034228.1 | NP_001029400.1 | 4 - 5 | 359 | 70 | |
| GenBank mRNA | BC103130.1 | - | 4 - 5 | 359 | 70 | |
More Information
Set Membership: |
|

EIF4A1
Genomic Map