Assay Details
Gene Symbol: MED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034237.2 | NP_001029409.1 | 6 - 7 | 602 | 85 | |
GenBank mRNA | BC102648.1 | - | 6 - 7 | 603 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
