Assay Details
Gene Symbol: ANAPC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034243.1 | NP_001029415.1 | 4 - 5 | 356 | 127 | |
GenBank mRNA | BC103197.1 | - | 4 - 5 | 356 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
