Assay Details
| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034256.2 | NP_001029428.1 | 5 - 6 | 438 | 85 | |
| GenBank mRNA | BC102530.1 | - | 5 - 6 | 439 | 85 | |
| BT026526.1 | - | 3 - 4 | 610 | 85 | ||
More Information
Set Membership: |
|

COA1
Genomic Map