Assay Details
| Gene Symbol: ARL6IP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034281.2 | NP_001029453.1 | 3 - 4 | 594 | 129 | |
| GenBank mRNA | BC105178.1 | - | 3 - 4 | 597 | 129 | |
More Information
Set Membership: |
|

ARL6IP6
Genomic Map