Assay Details
Gene Symbol: MPST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034291.2 | NP_001029463.2 | 1 - 2 | 101 | 72 | |
GenBank mRNA | BC105248.1 | - | 1 - 2 | 68 | 72 | |
DT812437.1 | - | 1 - 2 | 101 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC105248.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
