Assay Details
| Gene Symbol: NIPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034298.1 | NP_001029470.1 | 7 - 8 | 720 | 90 | |
| GenBank mRNA | BC104627.1 | - | 7 - 8 | 720 | 90 | |
More Information
Set Membership: |
|

NIPA2
Genomic Map