Assay Details
| Gene Symbol: PGK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034299.1 | NP_001029471.1 | 4 - 5 | 535 | 73 | |
| GenBank mRNA | BC102308.1 | - | 4 - 5 | 535 | 73 | |
| BT021601.1 | - | 4 - 5 | 465 | 73 | ||
More Information
Set Membership: |
|

PGK1
Genomic Map