Assay Details
Gene Symbol: SPARCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034302.2 | NP_001029474.1 | 6 - 7 | 1620 | 70 | |
GenBank mRNA | BC103351.1 | - | 6 - 7 | 1621 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
