Assay Details
| Gene Symbol: CCDC28B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034305.2 | NP_001029477.1 | 2 - 3 | 250 | 76 | |
| GenBank mRNA | BC102209.1 | - | 2 - 3 | 251 | 76 | |
More Information
Set Membership: |
|

CCDC28B
Genomic Map