Assay Details
| Gene Symbol: OSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034328.2 | NP_001029500.1 | 2 - 3 | 1148 | 64 | |
| GenBank mRNA | BC102143.1 | - | 2 - 3 | 1149 | 64 | |
More Information
Set Membership: |
|

OSR2
Genomic Map