Assay Details
| Gene Symbol: PERP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034350.2 | NP_001029522.1 | 2 - 3 | 416 | 99 | |
| GenBank mRNA | BC102419.1 | - | 2 - 3 | 417 | 99 | |
More Information
Set Membership: |
|

PERP
Genomic Map