Assay Details
| Gene Symbol: APOBEC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034355.1 | NP_001029527.1 | 1 - 2 | 158 | 76 | |
| GenBank mRNA | BC103438.1 | - | 1 - 2 | 158 | 76 | |
| DQ280381.1 | - | 1 - 2 | 174 | 76 | ||
More Information
Set Membership: |
|

APOBEC2
Genomic Map