Assay Details
| Gene Symbol: APOBEC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034355.1 | NP_001029527.1 | 2 - 3 | 725 | 72 | |
| GenBank mRNA | BC103438.1 | - | 2 - 3 | 725 | 72 | |
| DQ280381.1 | - | 2 - 3 | 741 | 72 | ||
More Information
Set Membership: |
|

APOBEC2
Genomic Map