Assay Details
| Gene Symbol: GET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034359.2 | NP_001029531.1 | 3 - 4 | 343 | 88 | |
| GenBank mRNA | BC103216.1 | - | 3 - 4 | 344 | 88 | |
More Information
Set Membership: |
|

GET1
Genomic Map