Assay Details
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034373.2 | NP_001029545.1 | 3 - 4 | 736 | 91 | |
GenBank mRNA | BC105150.1 | - | 3 - 4 | 738 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
