Assay Details
| Gene Symbol: CYP4V2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034373.2 | NP_001029545.1 | 4 - 5 | 926 | 124 | |
| GenBank mRNA | BC105150.1 | - | 4 - 5 | 928 | 124 | |
More Information
Set Membership: |
|

CYP4V2
Genomic Map