Assay Details
Gene Symbol: CTSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034385.2 | NP_001029557.1 | 9 - 10 | 739 | 70 | |
GenBank mRNA | BC102386.1 | - | 9 - 10 | 740 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
