Assay Details
Gene Symbol: C1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034407.1 | NP_001029579.1 | 4 - 5 | 632 | 58 | |
GenBank mRNA | BC103405.1 | - | 4 - 5 | 632 | 58 | |
BT030568.1 | - | 4 - 5 | 615 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
