Assay Details
| Gene Symbol: RAB5IF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034421.2 | NP_001029593.1 | 3 - 4 | 527 | 76 | |
| GenBank mRNA | BC102069.1 | - | 3 - 4 | 528 | 76 | |
More Information
Set Membership: |
|

RAB5IF
Genomic Map