Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034434.2 | NP_001029606.1 | 3 - 4 | 234 | 80 | |
| GenBank mRNA | BC102445.1 | - | 3 - 4 | 235 | 80 | |
More Information
Set Membership: |
|

RPL30
Genomic Map