Assay Details
Gene Symbol: PSMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034441.2 | NP_001029613.1 | 2 - 3 | 254 | 59 | |
GenBank mRNA | BC103247.1 | - | 2 - 3 | 257 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
