Assay Details
| Gene Symbol: DYNLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034442.2 | NP_001029614.1 | 2 - 3 | 131 | 61 | |
| GenBank mRNA | BC102138.1 | - | 2 - 3 | 133 | 61 | |
More Information
Set Membership: |
|

DYNLRB1
Genomic Map