Assay Details
| Gene Symbol: RPL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034459.2 | NP_001029631.1 | 1 | 548 | 80 | |
| GenBank mRNA | BC102600.1 | - | 1 | 551 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL17| Gene Symbol: RPL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034459.2 | NP_001029631.1 | 1 | 548 | 80 | |
| GenBank mRNA | BC102600.1 | - | 1 | 551 | 80 | |
Set Membership: |
|