Assay Details
Gene Symbol: MESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034469.1 | NP_001029641.1 | 1 - 2 | 232 | 60 | |
GenBank mRNA | BC102262.1 | - | 1 - 2 | 232 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
