Assay Details
Gene Symbol: MED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034486.1 | NP_001029658.1 | 1 - 2 | 129 | 71 | |
GenBank mRNA | BC103312.1 | - | 1 - 2 | 129 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
