Assay Details
| Gene Symbol: EIF3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034489.2 | NP_001029661.1 | 1 - 2 | 138 | 57 | |
| GenBank mRNA | BC102250.1 | - | 1 - 2 | 139 | 57 | |
More Information
Set Membership: |
|

EIF3K
Genomic Map