Assay Details
Gene Symbol: MRPL58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034493.1 | NP_001029665.1 | 4 - 5 | 388 | 104 | |
GenBank mRNA | BC102162.1 | - | 4 - 5 | 388 | 104 | |
BC134736.1 | - | 4 - 5 | 301 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
