Assay Details
| Gene Symbol: MRPL58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034493.1 | NP_001029665.1 | 5 - 6 | 559 | 74 | |
| GenBank mRNA | BC102162.1 | - | 5 - 6 | 559 | 74 | |
| BC134736.1 | - | 5 - 6 | 472 | 74 | ||
More Information
Set Membership: |
|

MRPL58
Genomic Map