Assay Details
| Gene Symbol: RPL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034495.1 | NP_001029667.1 | 3 - 4 | 262 | 86 | |
| GenBank mRNA | BC105179.1 | - | 3 - 4 | 262 | 86 | |
More Information
Set Membership: |
|

RPL35
Genomic Map