Assay Details
| Gene Symbol: CBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034513.1 | NP_001029685.1 | 1 - 2 | 396 | 77 | |
| GenBank mRNA | BC102943.1 | - | 1 - 2 | 396 | 77 | |
More Information
Set Membership: |
|

CBR1
Genomic Map