Assay Details
| Gene Symbol: MRPS26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034514.2 | NP_001029686.1 | 3 - 4 | 506 | 73 | |
| GenBank mRNA | BC103054.1 | - | 3 - 4 | 507 | 73 | |
More Information
Set Membership: |
|

MRPS26
Genomic Map