Assay Details
| Gene Symbol: GPRC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034515.1 | NP_001029687.1 | 1 - 2 | 130 | 85 | |
| GenBank mRNA | BC105147.1 | - | 1 - 2 | 130 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105147.1, NM_001034515.1 transcripts.
Set Membership: |
|

GPRC5A
Genomic Map