Assay Details
| Gene Symbol: PEX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034540.2 | NP_001029712.1 | 1 - 2 | 88 | 61 | |
| GenBank mRNA | BC102949.1 | - | 1 - 2 | 89 | 61 | |
More Information
Set Membership: |
|

PEX19
Genomic Map