Assay Details
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034550.2 | NP_001029722.1 | 2 - 3 | 341 | 65 | |
GenBank mRNA | BC102258.1 | - | 2 - 3 | 342 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
