Assay Details
| Gene Symbol: ACOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034560.1 | NP_001029732.1 | 7 - 8 | 680 | 73 | |
| GenBank mRNA | BC104625.1 | - | 7 - 8 | 680 | 73 | |
| BT030514.1 | - | 7 - 8 | 726 | 73 | ||
More Information
Set Membership: |
|

ACOT9
Genomic Map