Assay Details
| Gene Symbol: DDX19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034571.2 | NP_001029743.1 | 2 - 3 | 222 | 99 | |
| GenBank mRNA | BC103093.1 | - | 2 - 3 | 223 | 99 | |
| BT021652.1 | - | 2 - 3 | 194 | 99 | ||
More Information
Set Membership: |
|

DDX19A
Genomic Map