Assay Details
| Gene Symbol: MRPL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034576.2 | NP_001029748.1 | 2 - 3 | 274 | 67 | |
| GenBank mRNA | BC103424.1 | - | 2 - 3 | 275 | 67 | |
More Information
Set Membership: |
|

MRPL24
Genomic Map