Assay Details
| Gene Symbol: OVCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034601.2 | NP_001029773.1 | 1 - 2 | 217 | 63 | |
| GenBank mRNA | BC103277.1 | - | 1 - 2 | 218 | 63 | |
More Information
Set Membership: |
|

OVCA2
Genomic Map