Assay Details
| Gene Symbol: EIF3CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034618.2 | NP_001029790.1 | 9 - 10 | 1018 | 76 | |
| GenBank mRNA | BC103352.1 | - | 9 - 10 | 1019 | 76 | |
More Information
Set Membership: |
|

EIF3CL
Genomic Map