Assay Details
| Gene Symbol: CCNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034650.2 | NP_001029822.2 | 8 - 9 | 1058 | 68 | |
| GenBank mRNA | BC104532.1 | - | 3 - 4 | 1009 | 68 | |
| DT814268.1 | - | 3 - 4 | 325 | 68 | ||
| EE332955.1 | - | 2 - 3 | 195 | 68 | ||
More Information
Set Membership: |
|

CCNL2
Genomic Map