Assay Details
| Gene Symbol: ZCCHC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034652.2 | NP_001029824.1 | 7 - 8 | 599 | 67 | |
| GenBank mRNA | BC102395.1 | - | 7 - 8 | 601 | 67 | |
| BT025465.1 | - | 6 - 7 | 494 | 67 | ||
More Information
Set Membership: |
|

ZCCHC17
Genomic Map