Assay Details
| Gene Symbol: GDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034686.2 | NP_001029858.1 | 5 - 6 | 926 | 82 | |
| GenBank mRNA | BC102273.1 | - | 5 - 6 | 927 | 82 | |
More Information
Set Membership: |
|

GDE1
Genomic Map