Assay Details
| Gene Symbol: COX7A2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034695.2 | NP_001029867.1 | 1 - 2 | 118 | 59 | |
| GenBank mRNA | BC102552.1 | - | 1 - 2 | 119 | 59 | |
More Information
Set Membership: |
|

COX7A2L
Genomic Map