Assay Details
| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034698.2 | NP_001029870.1 | 5 - 6 | 1222 | 58 | |
| GenBank mRNA | BC102747.1 | - | 5 - 6 | 1223 | 58 | |
More Information
Set Membership: |
|

SERPINC1
Genomic Map